Silica gel, TLC high purity grade, without binder, Alfa Aesar™

For TLC Plate Preparation

Manufacturer: Alfa Aesar 4275618

Quantity 50g

 View more versions of this product

Catalog No. AA4275618

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 63231-67-4
    Physical Form Powder
    MDL Number MFCD00011232
    Chemical Name or Material Silica gel
    Grade TLC High Purity Grade

    Specifications

    Physical Form Powder
    Quantity 50g
    Color White
    EINECS Number 231-545-4
    Odor Odorless
    MDL Number MFCD00011232
    CAS 63231-67-4
    Health Hazard 1 H373: May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Silica gel
    Grade TLC High Purity Grade

    H373: May cause damage to organs through prolonged or repeated exposure.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit