Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silica Standard, 1mL = 1mg SiO2, Ricca Chemical
Click to view available options
Quantity:
120mL
20L
Packaging:
Cubitainer
Poly natural
Specifications
CAS Min % | 97.0,0.51 |
CAS Max % | 100.0,0.53 |
Physical Form | Liquid |
Packaging | Poly natural |
Equivalence | 1 mL = 1 mg SiO |
Color | Colorless |
Quantity | 120mL |
Chemical Name or Material | Silica Standard |