Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silica Standard Solution, APHA, Spectrum™ Chemical
Specifications
Concentration | 1.000 ± 0.010 mg SiO₂/mL |
---|---|
Description | CAS: 13517-24-3; 7732-18-5 |
Packaging | Poly Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18-613-101
|
Spectrum Chemical Manufacturing Corporation
S135125ML |
125 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Specifications
1.000 ± 0.010 mg SiO₂/mL | |
CAS: 13517-24-3; 7732-18-5 | |
Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title