Learn More
Trajan™ Non-Deactivated Fused Silica GC Tubing
Used in a wide range of capillary GC, LC and Bioanalytical applications. Trajan Scientific and Medical™ Non-Deactivated Fused Silica GC Tubing is protected with a high temperature Polyimide resin
Specifications
Deactivation | Not deactivated |
---|---|
For Use With (Application) | GC, LC and Bioanalytical Applications |
Material | Silica |
Description
- Used in a wide range of capillary GC, LC and Bioanalytical applications
- Made from high quality fused silica
- Tubing protected with a high temperature Polyimide resin
- 0.005mm-0.53mm internal diameters
Specifications
Not deactivated | |
GC, LC and Bioanalytical Applications | |
Silica |