Learn More
Silicagel, functionalized, 4-Bromophenyl, ca. 1 mmol/g, part. size: 40-63 micron, ACROS Organics
Manufacturer: Acros Organics 376480010
Description
For solid support synthesisSpecifications
Silica gel, functionalized, 4-bromophenyl | |
7631-86-9, 108-86-1 | |
Glass bottle | |
1g | |
O2Si | |
15, 8632 | |
Solubility in water: insoluble. | |
Crystalline Powder |
Functionalized, 4-Bromophenyl, ca. 1 mmol/g | |
(molecular) ca. 1mmol/g | |
40 to 63 Micron | |
White | |
MFCD03096006 | |
4-Bromophenyl substituted silicagel | |
60.08 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.