Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicagel, functionalized, 4-Bromophenyl, ca. 1 mmol/g, part. size: 40-63 micron, ACROS Organics™

Manufacturer:  Acros Organics 376480050

 View more versions of this product

Catalog No. AC376480050


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

For solid support synthesis
Specifications

Specifications

Silica gel, functionalized, 4-bromophenyl
7631-86-9, 108-86-1
Glass bottle
5g
O2Si
15, 8632
Solubility in water: insoluble.
Crystalline Powder
Functionalized, 4-Bromophenyl, ca. 1 mmol/g
(molecular) ca. 1mmol/g
40 to 63 Micron
White
MFCD03096006
4-Bromophenyl substituted silicagel
60.08
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit