Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicic Acid, n-Hydrate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Poly Pail

Chemical Identifiers

63231-67-4
H2O3Si
78.098
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
dihydroxy(oxo)silane
O[Si](=O)O

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Physical Form Powder
Packaging Poly Pail
Specific Gravity 1L = 2.10kg
Chemical Name or Material Silicic Acid, n-Hydrate
Grade BAKER ANALYZED™ Reagent