Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon, 99+%, powder, -325 mesh, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass Bottle
7440-21-3
Si
28.085
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]
7440-21-3
Si
28.085
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
5461123
CHEBI:27573
silicon
[Si]
Assay Percent Range 99+%
Merck Index 15, 8630
Formula Weight 28.09
Physical Form Fine Powder
Percent Purity 99+%
Packaging Glass Bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in molten alklai oxides, soluble in a mixture of nitric acid and hydrofluor, alkalis, insoluble in nitric acid and hydrochloric acid
Total Trace Metal Impurities 1.0% max.
Boiling Point 2355.0°C
Color Gray
Melting Point 1410.0°C
Quantity 10g
Chemical Name or Material Silicon, -325 mesh
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning