Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon, AAS standard solution, Si 1000µg/ml, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 88095AE

Quantity 100mL

 View more versions of this product

Catalog No. AA88095AE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

Liquid
Si
MFCD00085311
Silicon, AAS standard solution, AAS Standard Solution

Specifications

Liquid
Si
Nitric acid: 5%; Hydrogen fluoride: 0.1%; Silicon: 0.1%; Water: 94.8%
100mL
Liquid
MFCD00085311
23 hPa (17mm Hg) at 20°C
Silicon, AAS standard solution, AAS Standard Solution

Safety and Handling

Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.

DOTInformation : DOT Class: 8, Packaging Group: III

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit