Alfa Aesar™ Silicon carbide, beta-phase

Manufacturer: Alfa Aesar™ 4464722

Quantity 100g

 View more versions of this product

Catalog No. AA4464722

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 409-21-2
    Molecular Formula SiC
    Formula Weight 40.1
    MDL Number MFCD00049531
    Merck Index 14,8492
    Chemical Name or Material Silicon carbide, beta-phase

    Specifications

    Formula Weight 40.1
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula SiC
    Odor Odorless
    Particle Size 0.1 to 0.2 Micron APS
    CAS 409-21-2
    Quantity 100g
    Surface Area 20 to 40m2/g
    Merck Index 14,8492
    Melting Point 2700°C
    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00049531
    Health Hazard 2 GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Silicon carbide, beta-phase

    GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit