Learn More
Silicon carbide, beta-phase, 99% (metals basis), Thermo Scientific Chemicals
Description
Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2700°C |
Color | Black to Gray |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.8% (metals basis) |
Solubility Information | Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron. |
Formula Weight | 40.1 |
Odor | Odorless |
Chemical Name or Material | Silicon carbide, beta-phase |
Safety and Handling
Hazard Category | H351-H315-H319-H335: Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
EINECSNumber | 206-991-8 |
RTECSNumber | VW0450000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.