Silicon carbide, beta-phase, 99.8% (metals basis), Alfa Aesar™

S.A. 11.5 m2/g

Manufacturer: Alfa Aesar 1416522

Quantity 100g

 View more versions of this product

Catalog No. AA1416522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 409-21-2
    Physical Form Powder
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula SiC
    Formula Weight 40.1
    MDL Number MFCD00049531
    Chemical Name or Material Silicon carbide, beta-phase

    Specifications

    Physical Form Powder
    Formula Weight 40.1
    Quantity 100g
    Color Gray to black
    EINECS Number 206-991-8
    Odor Odorless
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula SiC
    MDL Number MFCD00049531
    CAS 409-21-2
    Health Hazard 1 H351-H315-H319-H335: Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Silicon carbide, beta-phase

    H351-H315-H319-H335: Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit