Alfa Aesar™ Silicon carbide, beta-phase, nanopowder, 95% min

Manufacturer: Alfa Aesar™ 4388422

Quantity 100g

 View more versions of this product

Catalog No. AA4388422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 409-21-2
    Molecular Formula SiC
    Formula Weight 40.1
    MDL Number MFCD00049531
    Merck Index 14,8492
    Chemical Name or Material Silicon carbide, beta-phase

    Specifications

    Formula Weight 40.1
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Surface Area >80m2/g
    Merck Index 14,8492
    Color Gray to black
    Solubility Information Insoluble in water, alcohol; Soluble in fused alkali
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number VW0450000
    Odor Odorless
    Physical Form Nanopowder
    CAS 409-21-2
    TSCA TSCA
    Particle Size <30nm APS
    Molecular Formula SiC
    MDL Number MFCD00049531
    Melting Point 2700°C
    Quantity 100g
    Chemical Name or Material Silicon carbide, beta-phase
    Assay ≥95%

    GHS H Statement
    H351-H315-H319-H335
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit