Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon Dioxide, 325 Mesh, Crystalline, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
14808-60-7
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O
14808-60-7
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O

SI108, 14808-60-7, SiO2

Spectrum™ Silicon Dioxide, 325 Mesh, Crystalline is also referred to as silica. Found naturally under the form of quartz and in some living organism, it is one of the most complex and abundant families of materials. Nearly 95% of all silicon dioxide is used in the construction industry in the form of glass and Portland cement among others. It is also used as a powder flow helper in the production of tablets.

CAS Max % 1
Quantity 45 kg
Packaging Fiber Drum
Grade Reagent