Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon Dioxide, 325 Mesh, Crystalline, Spectrum™ Chemical
SDP

Catalog No. 18606212
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle

SI108, 14808-60-7, SiO2

Spectrum™ Chemical Silicon Dioxide, 325 Mesh, Crystalline is also referred to as silica. Found naturally under the form of quartz and in some living organism, it is one of the most complex and abundant families of materials. Nearly 95% of all silicon dioxide is used in the construction industry in the form of glass and Portland cement among others. It is also used as a powder flow helper in the production of tablets.

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Packaging Poly Bottle
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.