Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silicon granular, virgin poly fines, 99.999% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 500 g |
Percent Purity | 99.999% |
Odor | Odorless |
Particle Size | ∽1/4 in. & down |
Form | Chunks to Powder |
Assay | (metals basis) |
Chemical Name or Material | Silicon granular, Virgin poly fines |
Safety and Handling
missing translation for 'healthHazard1' | Warning |
missing translation for 'healthHazard2' | GHS H Statement H319 Causes serious eye irritation. |
missing translation for 'healthHazard3' | GHS P Statement P280-P264-P305+P351+P338-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. Wash thoroughly after handling. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
EINECSNumber | 231-130-8 |
RTECSNumber | VW0400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'