Alfa Aesar™ Silicon(II) oxide, 99.9% (metals basis)

For vacuum deposition

Manufacturer: Alfa Aesar™ 4171136

Quantity 500g

 View more versions of this product

Catalog No. AA4171136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10097-28-6
    Formula Weight 44.09
    MDL Number MFCD00151536
    Merck Index 14,8496
    Synonym Oxosilylene
    Chemical Name or Material Silicon(II) oxide, 6 to 15mm Pieces

    Specifications

    Formula Weight 44.09
    Physical Form Pieces
    MDL Number MFCD00151536
    Merck Index 14,8496
    Boiling Point 1880°C
    TSCA TSCA
    Synonym Oxosilylene
    Melting Point >1702°C
    Quantity 500g
    Density 2.13g/mL
    Odor Odorless
    CAS 10097-28-6
    Chemical Name or Material Silicon(II) oxide, 6 to 15mm Pieces
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit