Silicon(II) oxide, Optical Grade, 99.8% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 8943018

Quantity 50g

 View more versions of this product

Catalog No. AA8943018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10097-28-6
    Physical Form Powder
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula OSi
    Formula Weight 44.09
    MDL Number MFCD00151536
    Synonym Oxosilylene
    Chemical Name or Material Silicon(II) oxide
    Grade Optical Grade

    Specifications

    Physical Form Powder
    Formula Weight 44.09
    Quantity 50g
    Color Black
    EINECS Number 233-232-8
    Odor Odorless
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula OSi
    MDL Number MFCD00151536
    CAS 10097-28-6
    Synonym Oxosilylene
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Silicon(II) oxide
    Grade Optical Grade

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit