Alfa Aesar™ Silicon(II) oxide, Optical Grade, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 6916614

Quantity 25g

 View more versions of this product

Catalog No. AA6916614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 10097-28-6
    Molecular Formula OSi
    Linear Formula SiO
    Formula Weight 44.09
    MDL Number MFCD00151536
    Merck Index 14,8496
    Synonym Oxosilylene
    Chemical Name or Material Silicon (II) oxide
    Grade Optical Grade

    Specifications

    Formula Weight 44.09
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 10097-28-6
    MDL Number MFCD00151536
    Merck Index 14,8496
    Boiling Point 1880°C
    Mesh Size −3/8 in. +10 Mesh
    Molecular Formula OSi
    Melting Point >1702°C
    Health Hazard 1 Warning
    TSCA TSCA
    Odor Odorless
    Linear Formula SiO
    Synonym Oxosilylene
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Density 2.13g/mL
    Chemical Name or Material Silicon (II) oxide
    Assay 99.9%, (metals basis)
    Grade Optical Grade

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit