Alfa Aesar™ Silicon(II) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.9% (metals basis)

Magnetron sputtering source

Manufacturer: Alfa Aesar™ 41092KS

Quantity 1 each

 View more versions of this product

Catalog No. AA41092KS

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Formula Weight 44.09
    MDL Number MFCD00151536
    Synonym Oxosilylene
    Chemical Name or Material Silicon(II) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick

    Specifications

    Formula Weight 44.09
    MDL Number MFCD00151536
    TSCA TSCA
    Synonym Oxosilylene
    Quantity 1 each
    Chemical Name or Material Silicon(II) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit