Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) chloride, 99.8+%, ACROS Organics™

missing translation for 'orderingAttributeHoverText'
Quantity:
50mL
500mL
2.5L
Packaging:
Glass bottle
10026-04-7
Cl4Si
169.9
MFCD00011229
FDNAPBUWERUEDA-UHFFFAOYSA-N
silicon tetrachloride,silicon chloride,perchlorosilane,silane, tetrachloro,silicon iv chloride,tetrachlorosilicon,chlorid kremicity,silicio tetracloruro di,siliciumtetrachlorid,silicon chloride sicl4
24816
tetrachlorosilane
[Si](Cl)(Cl)(Cl)Cl
10026-04-7
Cl4Si
169.9
MFCD00011229
FDNAPBUWERUEDA-UHFFFAOYSA-N
silicon tetrachloride,silicon chloride,perchlorosilane,silane, tetrachloro,silicon iv chloride,tetrachlorosilicon,chlorid kremicity,silicio tetracloruro di,siliciumtetrachlorid,silicon chloride sicl4
24816
tetrachlorosilane
[Si](Cl)(Cl)(Cl)Cl
Aluminum (Al) 5ppm max.
Assay Percent Range 99.8+%
Linear Formula SiCl4
Antimony (Sb) 0.1ppm max.
Fieser 10,347; 11,466; 16,317
Merck Index 15, 8639
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with benzene,ether,chloroform,,petroleum ether,carbon tetrachloride,,tin tetrachloride,sulfur,mono- and dichlorides,decomposes by alcohol with evolution of hydrogen,chloride
Formula Weight 169.9
Physical Form Liquid
Percent Purity 99.8+%
Chromium (Cr) 0.2ppm max.
Cobalt (Co) 0.1ppm max.
Copper (Cu) 0.1ppm max.
Density 1.4800g/mL
Iron (Fe) 1ppm max.
Manganese (Mn) 0.1ppm max.
Nickel (Ni) 0.1ppm max.
Molybdenum (Mo) 0.1ppm max.
Packaging Glass bottle
Purity 99.8% min
Refractive Index 1.413
Solubility Solubility in water: decomposes. Other solubilities: miscible with benzene, ether, chloroform,, petroleum ether, carbon tetrachloride,, tin tetrachloride, sulfur, mono- and dichlorides, decomposes by alcohol with evolution of hydrogen, chloride
Specific Gravity 1.48
Storage Information Product may develop some turbidity or precipitate
Titanium (Ti) 1ppm max.
Trace Analysis 0.1ppm max.
Boiling Point 58.0°C
Color Colorless
Melting Point -70.0°C
Quantity 500mL
Chemical Name or Material Silicon(IV) chloride, 99.8+%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Danger

EINECSNumber : 233-054-0

RTECSNumber : VW0525000

TSCA : TSCA