Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) iodide, 99% (metals basis), Alfa Aesar™

Light, Air and Moisture Sensitive

Manufacturer:  Alfa Aesar 8730303

Quantity 1g

 View more versions of this product

Catalog No. AA8730303

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13465-84-4
99% (metals basis)
535.7
Silicon tetraiodide
Crystals, Packaged Under Argon
I4Si
MFCD00054041
Silicon(IV) iodide

Specifications

13465-84-4
99% (metals basis)
535.7
Silicon tetraiodide
1g
Crystals, Packaged Under Argon
I4Si
MFCD00054041
White
Silicon(IV) iodide

Safety and Handling

SDS & Safety Documents

H300-H314-H318: Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 236-706-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit