Alfa Aesar™ Silicon(IV) nitride, 98% min

Manufacturer: Alfa Aesar™ 4388522

Quantity 100g

 View more versions of this product

Catalog No. AA4388522

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 12033-89-5
    Molecular Formula Si3N4
    Formula Weight 140.28
    MDL Number MFCD00011230
    Chemical Name or Material Silicon (IV) nitride

    Specifications

    Formula Weight 140.28
    Physical Form Powder
    Molecular Formula Si3N4
    CAS 12033-89-5
    Particle Size 30nm
    MDL Number MFCD00011230
    Melting Point 1900°C
    TSCA TSCA
    Odor Odorless
    Surface Area 109m2/g
    Quantity 100g
    Chemical Name or Material Silicon (IV) nitride
    Assay ≥98%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit