Alfa Aesar™ Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase

Manufacturer: Alfa Aesar™ 4037909

Quantity 10g

 View more versions of this product

Catalog No. AA4037909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12033-89-5
    Linear Formula Si3N4
    Formula Weight 140.28
    MDL Number MFCD00011230
    Merck Index 14,8497
    Chemical Name or Material Silicon(IV) nitride, Typically 90% beta-phase

    Specifications

    Formula Weight 140.28
    Mesh Size −325 Mesh
    Physical Form Powder
    Linear Formula Si3N4
    Merck Index 14,8497
    Color Gray-black
    Melting Point 1900°C (subl.)
    Quantity 10g
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00011230
    CAS 12033-89-5
    Density 3.2g/mL
    RTECS Number VW0650000
    Chemical Name or Material Silicon(IV) nitride, Typically 90% beta-phase
    Assay 99.3%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit