Alfa Aesar™ Silicon(IV) nitride, amorphous, nanopowder, 98.5+%

Manufacturer: Alfa Aesar™ 4561114

Quantity 25g

 View more versions of this product

Catalog No. AA4561114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12033-89-5
    Molecular Formula Si3N4
    Formula Weight 140.28
    MDL Number MFCD00011230
    Merck Index 14,8497
    Chemical Name or Material Silicon (IV) nitride, Amorphous

    Specifications

    Formula Weight 140.28
    TSCA TSCA
    Molecular Formula Si3N4
    CAS 12033-89-5
    Merck Index 14,8497
    Color Pale gray
    Melting Point 1900°C (subl.)
    Particle Size 15 to 30nm APS
    Physical Form Nanopowder
    MDL Number MFCD00011230
    RTECS Number VW0650000
    Odor Odorless
    Surface Area 103 to 123m2/g
    Quantity 25g
    Density 3.2g/mL
    Chemical Name or Material Silicon (IV) nitride, Amorphous
    Assay ≥98.5%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit