Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) nitride, MgO binder, sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA41088KS
Click to view available options
Quantity:
1 Ea.
MFCD00011230
MFCD00011230

 

Magnetron sputtering source

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
Formula Weight 140.28
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silicon(IV) nitride, MgO binder, Sputtering Target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
EINECSNumber 234-796-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit