Alfa Aesar™ Silicon(IV) oxide, 30% in Ethylene glycol, colloidal dispersion

Manufacturer: Alfa Aesar™ 43093A7

Quantity 5kg

 View more versions of this product

Catalog No. AA43093A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-86-9
    Molecular Formula O2Si
    Linear Formula SiO2
    Formula Weight 60.09
    MDL Number MFCD00011232
    Chemical Name or Material Silicon (IV) oxide, Colloidal dispersion

    Specifications

    Physical Form Particles In Liquid
    Formula Weight 60.09
    Particle Size 0.05 Micron
    CAS 7631-86-9
    Odor Sweet
    MDL Number MFCD00011232
    Color White
    Molecular Formula O2Si
    Linear Formula SiO2
    Quantity 5kg
    Concentration 30% in Ethylene glycol
    Density 1.3g/mL
    TSCA TSCA
    Chemical Name or Material Silicon (IV) oxide, Colloidal dispersion

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit