Silicon(IV) oxide, 30% in H2O, colloidal dispersion, Alfa Aesar™

For Use As a Binder in Ceramic Investment Shell Coating, With Refractory Powders For Yielding Superior Face Coats

Manufacturer: Alfa Aesar 43111C4

Quantity 25kg

 View more versions of this product

Catalog No. AA43111C4

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7631-86-9
    Physical Form Liquid
    Molecular Formula SiO2
    Formula Weight 60.09
    MDL Number MFCD00011232
    Chemical Name or Material Silicon(IV) oxide, 30% in H2O

    Specifications

    Formula Weight 60.09
    Quantity 25kg
    Color Colorless
    EINECS Number 231-545-4
    Physical Form Liquid
    Molecular Formula SiO2
    MDL Number MFCD00011232
    CAS 7631-86-9
    Chemical Name or Material Silicon(IV) oxide, 30% in H2O

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit