Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon(IV) oxide, 40% in H2O, 0.014 micron particles in colloidal dispersion, Alfa Aesar™

S.A. 250 m2/g

Manufacturer:  Alfa Aesar 43115A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43115A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7631-86-9
SiO2
MFCD00011232
Liquid
60.09
Silicon(IV) oxide, 40% in H2O

Specifications

7631-86-9
SiO2
MFCD00011232
Odorless
1kg
Liquid
60.09
1.30g/cm3 at 20°C
Colorless
Silicon(IV) oxide, 40% in H2O

Safety and Handling

SDS & Safety Documents

EINECSNumber : 231-545-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit