Silicon(IV) oxide, 40% in H2O, 0.02 micron particles in colloidal dispersion, Alfa Aesar™

S.A. 200 m2/g

Manufacturer: Alfa Aesar 43110A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43110A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-86-9
    Physical Form Liquid
    Molecular Formula SiO2
    Formula Weight 60.09
    MDL Number MFCD00011232
    Chemical Name or Material Silicon(IV) oxide, 40% in H2O

    Specifications

    Formula Weight 60.09
    Quantity 1kg
    Color Colorless
    Density 1.30g/cm3 at 20°C
    EINECS Number 231-545-4
    Physical Form Liquid
    Odor Odorless
    Molecular Formula SiO2
    MDL Number MFCD00011232
    CAS 7631-86-9
    Chemical Name or Material Silicon(IV) oxide, 40% in H2O

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit