Learn More
Silicon(IV) oxide, 50% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Description
Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles, and many other uses. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Primarily as a flow agent in powdered foods, or to adsorb water in hygroscopic applications. It is the primary component of diatomaceous earth. Colloidal silica is also used as a wine, beer, and juice fining agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Particles in Liquid |
Quantity | 1 kg |
Solubility Information | Fully miscible in water. Soluble in alkalis. |
Formula Weight | 60.09 |
Odor | Odorless |
Density | 1.40 g/cm3 at 20°C |
Chemical Name or Material | Silicon(IV) oxide, 50% in H2O |
Safety and Handling
EINECSNumber | 231-545-4 |
RTECSNumber | VV7565000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.