Silicon(IV) oxide, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1085614

Quantity 25g

 View more versions of this product

Catalog No. AA1085614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14808-60-7
    Physical Form Powder or Granules
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula SiO2
    Formula Weight 60.09
    MDL Number MFCD00011232
    Chemical Name or Material Silicon(IV) oxide

    Specifications

    Physical Form Powder or Granules
    Formula Weight 60.09
    Quantity 25g
    Color White to pale brown
    EINECS Number 238-878-4
    Assay Percent Range 99.999% (metals basis)
    Odor Odorless
    Molecular Formula SiO2
    MDL Number MFCD00011232
    CAS 14808-60-7
    Health Hazard 1 H350-H373: May cause cancer. May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Silicon(IV) oxide

    H350-H373: May cause cancer. May cause damage to organs through prolonged or repeated exposure.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit