Alfa Aesar™ Silicon(IV) oxide, amorphous fumed, S.A. 130-170m2/g

Adhesives, sealants, silicones

Manufacturer: Alfa Aesar™ 4273918

Quantity 50g

 View more versions of this product

Catalog No. AA4273918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-86-9
    Molecular Formula O2Si
    Linear Formula SiO2
    Formula Weight 60.09
    MDL Number MFCD00011232
    Merck Index 14,8493
    Chemical Name or Material Silicon (IV) oxide, Amorphous fumed

    Specifications

    Formula Weight 60.09
    Linear Formula SiO2
    Density 2.2g/mL
    Odor Odorless
    Quantity 50g
    Mesh Size −325 Mesh
    Physical Form Powder
    MDL Number MFCD00011232
    CAS 7631-86-9
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.
    Health Hazard 3 GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Surface Area 130 to 170m2/g
    Color White
    Merck Index 14,8493
    Melting Point 1710°C
    Molecular Formula O2Si
    Boiling Point 2230°C
    Chemical Name or Material Silicon (IV) oxide, Amorphous fumed

    GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit