Silicon(IV) oxide, Amorphous fumed, Surface treated, S.A. 105 to 130m2/g, Alfa Aesar™

For Use in Coatings, Adhesives, Sealants

Manufacturer: Alfa Aesar 42784A1

Quantity 1kg

 View more versions of this product

Catalog No. AA42784A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-86-9
    Physical Form Powder
    Molecular Formula O2Si
    Formula Weight 60.09
    MDL Number MFCD00011232
    Chemical Name or Material Silicon(IV) oxide

    Specifications

    Formula Weight 60.09
    Quantity 1kg
    Color White
    EINECS Number 231-545-4
    Physical Form Powder
    Surface Area or Specific Surface S.A.: 105 to 130m2/g
    Molecular Formula O2Si
    MDL Number MFCD00011232
    CAS 7631-86-9
    Odor Odorless
    Health Hazard 1 H373: May cause damage to organs through prolonged or repeated exposure.
    Health Hazard 2 Wear suitable protective clothing.
    Chemical Name or Material Silicon(IV) oxide

    Wear suitable protective clothing.

    H373: May cause damage to organs through prolonged or repeated exposure.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit