Alfa Aesar™ Silicon(IV) oxide, amorphous, nanopowder, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 4478214

Quantity 25g

 View more versions of this product

Catalog No. AA4478214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-86-9
    Molecular Formula SiOx, (x=1.2 to 1.6)
    MDL Number MFCD00011232
    Merck Index 14,8493
    Chemical Name or Material Silicon (IV) oxide, Amorphous

    Specifications

    Molecular Formula SiOx, (x=1.2 to 1.6)
    TSCA TSCA
    Odor Odorless
    Particle Size 10 to 20nm APS
    MDL Number MFCD00011232
    CAS 7631-86-9
    Melting Point 1710°C
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 2.4g/mL
    Physical Form Nanopowder
    Merck Index 14,8493
    Boiling Point 2230°C
    pH 6
    Health Hazard 2 GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Silicon (IV) oxide, Amorphous
    Assay 99.5%, (metals basis)

    GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit