Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silicon(IV) oxide, amorphous, nanopowder, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 4478222

Quantity 100g

 View more versions of this product

Catalog No. AA4478222


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7631-86-9
MFCD00011232
Silicon (IV) oxide, Amorphous
SiOx, (x=1.2 to 1.6)
14,8493

Specifications

7631-86-9
SiOx, (x=1.2 to 1.6)
14,8493
Odorless
2230°C
6
Silicon (IV) oxide, Amorphous
Nanopowder
MFCD00011232
2.4g/mL
10 to 20nm APS
1710°C
100g
99.5%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit