Alfa Aesar™ Silicon oxide, catalyst support, low surface area

Manufacturer: Alfa Aesar™ 43861A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43861A1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00011232
    Chemical Name or Material Silicon oxide, catalyst support, Low surface area

    Specifications

    Physical Form Pellet
    MDL Number MFCD00011232
    Quantity 1kg
    TSCA TSCA
    Particle Size 1/8 in.
    Chemical Name or Material Silicon oxide, catalyst support, Low surface area

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit