Silicon plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 13814-AP

Quantity 500mL

 View more versions of this product

Catalog No. AA13814AP

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula Si in 5% HNO3/tr· HF
    MDL Number MFCD00085311
    Chemical Name or Material Silicon, plasma standard solution

    Specifications

    Quantity 500mL
    DOT Information DOT Class: 8, Packaging Group: III
    Physical Form Liquid
    Appearance Liquid
    Molecular Formula Si in 5% HNO3/tr· HF
    MDL Number MFCD00085311
    Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Hydrogen fluoride: 0.1%; Silicon: 0.1%; Water: 94.8%
    Health Hazard 1 H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Silicon, plasma standard solution

    H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit