Alfa Aesar™ Silicon powder, crystalline, -325 mesh, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 3566203

Quantity 1g

 View more versions of this product

Catalog No. AA3566203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-21-3
    MDL Number MFCD00085311
    UN Number UN1346
    Chemical Name or Material Silicon powder

    Specifications

    Physical Form Crystalline Powder
    Quantity 1g
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378c
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    RTECS Number VW0400000
    Odor Odorless
    MDL Number MFCD00085311
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS
    Mesh Size −325 Mesh
    Color White
    TSCA TSCA
    CAS 7440-21-3
    UN Number UN1346
    Chemical Name or Material Silicon powder
    Assay 99.999%, (metals basis)

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378c
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit