Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silicon powder, crystalline/amorphous, typically 1-20 micron, 99.9985% (metals basis)

Manufacturer:  Alfa Aesar™ 3871530

Quantity 250g

 View more versions of this product

Catalog No. AA3871530


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-21-3
MFCD00085311
UN1346
Silicon powder

Specifications

White
7440-21-3
Odorless
MFCD00085311
Silicon powder
250g
typically 1 to 20 Micron
Crystalline/Amorphous Powder
UN1346
99.9985%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378c
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS

RTECSNumber : VW0400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit