Alfa Aesar™ Silicon powder, crystalline/amorphous, typically 1-20 micron, 99.9985% (metals basis)

Manufacturer: Alfa Aesar™ 3871518

Quantity 50g

 View more versions of this product

Catalog No. AA3871518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-21-3
    MDL Number MFCD00085311
    UN Number UN1346
    Chemical Name or Material Silicon powder

    Specifications

    Physical Form Crystalline/Amorphous Powder
    Quantity 50g
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378c
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Odor Odorless
    TSCA TSCA
    MDL Number MFCD00085311
    UN Number UN1346
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS
    Particle Size typically 1 to 20 Micron
    RTECS Number VW0400000
    CAS 7440-21-3
    Color White
    Chemical Name or Material Silicon powder
    Assay 99.9985%, (metals basis)

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378c
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit