Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly Bottle
6834-92-0,7732-18-5
Na2O3Si
122.062
MFCD00085311
NTHWMYGWWRZVTN-UHFFFAOYSA-N
23266
CHEBI:60720
disodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[Na+].[Na+]
6834-92-0,7732-18-5
Na2O3Si
122.062
MFCD00085311
NTHWMYGWWRZVTN-UHFFFAOYSA-N
23266
CHEBI:60720
disodium;dioxido(oxo)silane
[O-][Si](=O)[O-].[Na+].[Na+]

For atomic absorption. (1mL = 1mg Si) Solute: Sodium Silicate Solvent: Distilled Water

Concentration 990 to 1010ppm
Physical Form Liquid
Grade Certified
Packaging Poly Bottle
pH 11.8
Color Colorless
Quantity 100mL
Chemical Name or Material Silicon, Reference Standard Solution, 1000ppm ±1%

DOTInformation : DOT Class 8, : Corrosive