Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicone Fluid, 500, Spectrum™

Click to view available options
Quantity:
1 kg
100 g
44 lbs.
440 lbs.
Packaging:
Amber Glass Bottle
Drum
Poly Drum
63148-62-9
0.00
*
63148-62-9
0.00
*

SI112, 63148-62-9

Spectrum™ Silicone Fluid, 500
Packaging Drum
Viscosity 500 cs
Quantity 440 lbs.
CAS Max % 1
Grade Reagent