Learn More
Sklar InstrumentsSupplier Diversity Partner Silicone Instrument Holder for Cassettes with Detachable Lid, Sklar, 10 1/2 in. Silicone Instrument Holder for Cassettes with Detachable Lid 3 Piece, Maximum 10 Instrument Configuration, Grey. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106539
Silicone Instrument Holder for Cassettes with Detachable Lid, Sklar, 10 1/2 in. Silicone Instrument Holder for Cassettes with Detachable Lid 3 Piece, Maximum 10 Instrument Configuration, Grey. Sklar's Silicone Instrument Holders are used for positioning instruments inside a cassette with a detachable lid used for cleaning, storage, or sterilization. The holders are repositionable to assist in storing different types and size instruments within a casette. They are available in a variety of colors and sold in sets of 3. This product is grey, has an overall length of 10-1/2 inches and holds approximately 10 instruments.
Catalog No. 50-143-7241