Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicone oil, high temperature, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass Bottle
63148-58-3
Poly(methylphenylsiloxane)
63148-58-3
Poly(methylphenylsiloxane)
Flash Point 316°C
Viscosity 125 mm2/s (25°C)
Quantity 2.5kg
Refractive Index 1.4940 to 1.5010
Packaging Glass Bottle
Density 1.0500g/mL
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble in water.
Chemical Name or Material Silicone oil