Learn More
Selleck Chemical LLCSupplier Diversity Partner Silodosin 10mg 160970-54-7 KAD 3213, KMD 3213  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S161310MG
Silodosin (KAD 3213, KMD 3213) is a highly selective α1A-adrenoceptor antagonist, used in treatment of benign prostatic hyperplasia.
Catalog No. 50-654-6