Learn More
Trajan Scientific and Medical™ SilTite™ Column Connectors
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Manufacturer:  Trajan Scientific and Medical™ 073233
 View more versions of this product
Catalog No. 07-323-3
Add to cart
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
SilTite Nuts for Shimadzu
Installation for Capillary Columns
Column Connectors