Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan Scientific and Medical™ Thermo Scientific Split / Splitless Tapered FAST FocusLiner

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Manufacturer:  Trajan Scientific and Medical™ 092115

 View more versions of this product

Catalog No. 09-211-5


Add to cart

Description

Description

    Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Thermo Scientific instruments
Specifications

Specifications

78.5 mm
2.3 mm
Finnigan 9001 and GCQ GCs, and Trace 1300, 1310
25/Pk.
6.3 mm
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit