Alfa Aesar™ Silver arsenate, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4422822

Quantity 100g

 View more versions of this product

Catalog No. AA4422822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13510-44-6
    Molecular Formula Ag3AsO4
    Formula Weight 462.53
    MDL Number MFCD00046163
    UN Number UN1557
    Chemical Name or Material Silver arsenate

    Specifications

    Physical Form Powder/Lump
    Formula Weight 462.53
    Health Hazard 3 GHS P Statement
    P261-P281-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Color Brick red
    Sensitivity Light Sensitive
    Molecular Formula Ag3AsO4
    MDL Number MFCD00046163
    UN Number UN1557
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H331-H350
    Toxic if swallowed.
    Toxic if inhaled.
    May cause cancer.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
    CAS 13510-44-6
    Chemical Name or Material Silver arsenate
    Assay 99.99%, (metals basis)

    GHS H Statement
    H301-H331-H350
    Toxic if swallowed.
    Toxic if inhaled.
    May cause cancer.

    GHS P Statement
    P261-P281-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit