Alfa Aesar™ Silver, brazing alloy, flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long

Manufacturer: Alfa Aesar™ 41031KJ

Quantity 5 pieces

 View more versions of this product

Catalog No. AA41031KJ

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00801120
    Chemical Name or Material Silver, brazing Alloy, Flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long, Tensile Strength: 79,000 psi

    Specifications

    Quantity 5 pieces
    TSCA TSCA
    MDL Number MFCD00801120
    Avg. Mol. Wt. or Mol. Wt. Range Ag:Cu:Zn:Sn; 56:22:17:5 wt% Nontoxic, cadmium free
    Chemical Name or Material Silver, brazing Alloy, Flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long, Tensile Strength: 79,000 psi

    Description

    • Low temperature alloy for use on ferrous and nonferrous material; good color match for stainless material

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit